Assay Details
| Gene Symbol: Fam120c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198105.2 | NP_932773.2 | 14 - 15 | 2993 | 67 | |
| GenBank mRNA | AK030477.1 | - | 14 - 15 | 3042 | 67 | |
| AK136727.1 | - | 14 - 15 | 2989 | 67 | ||
| AK156920.1 | - | 14 - 15 | 2994 | 67 | ||
| AY150023.1 | - | 14 - 15 | 2997 | 67 | ||
| BC137973.1 | - | 14 - 15 | 2993 | 8860992 | 67 | |
| BC171940.1 | - | 14 - 15 | 2993 | 9054226 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam120c
Genomic Map