Assay Details
| Gene Symbol: Fam120c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198105.2 | NP_932773.2 | 6 - 7 | 1353 | 92 | |
| GenBank mRNA | AK030477.1 | - | 6 - 7 | 1402 | 92 | |
| AK136727.1 | - | 6 - 7 | 1349 | 92 | ||
| AK156920.1 | - | 6 - 7 | 1354 | 92 | ||
| AY150023.1 | - | 6 - 7 | 1357 | 92 | ||
| AY150024.1 | - | 6 - 7 | 1357 | 92 | ||
| BC137973.1 | - | 6 - 7 | 1353 | 8860992 | 92 | |
| BC171940.1 | - | 6 - 7 | 1353 | 9054226 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam120c
Genomic Map