Assay Details
| Gene Symbol: Fam120c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198105.2 | NP_932773.2 | 10 - 11 | 2327 | 69 | |
| GenBank mRNA | AK030477.1 | - | 10 - 11 | 2376 | 69 | |
| AK136727.1 | - | 10 - 11 | 2323 | 69 | ||
| AK156920.1 | - | 10 - 11 | 2328 | 69 | ||
| AY150023.1 | - | 10 - 11 | 2331 | 69 | ||
| BC137973.1 | - | 10 - 11 | 2327 | 8860992 | 69 | |
| BC171940.1 | - | 10 - 11 | 2327 | 9054226 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam120c
Genomic Map