Assay Details
| Gene Symbol: Mroh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126487.1 | NP_001119959.1 | 21 - 22 | 3715 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh7| Gene Symbol: Mroh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126487.1 | NP_001119959.1 | 21 - 22 | 3715 | 74 | |
Set Membership: |
|