Assay Details
| Gene Symbol: Mroh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126487.1 | NP_001119959.1 | 9 - 10 | 1946 | 101 | |
| GenBank mRNA | AK134749.1 | - | 2 - 3 | 182 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mroh7
Genomic Map