| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400025442.mm.1 |
TC0400003207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025442.mm.1 |
NM_001126487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025442.mm.1 |
NM_001126487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025445.mm.1 |
TC0400003207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025445.mm.1 |
NM_001126487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025445.mm.1 |
NM_001126487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025444.mm.1 |
TC0400003207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025444.mm.1 |
NM_001126487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025444.mm.1 |
NM_001126487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025441.mm.1 |
TC0400003207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025441.mm.1 |
NM_001126487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025441.mm.1 |
NM_001126487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025443.mm.1 |
TC0400003207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025443.mm.1 |
NM_001126487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025443.mm.1 |
NM_001126487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025446.mm.1 |
TC0400003207.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025446.mm.1 |
NM_001126487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025446.mm.1 |
NM_001126487.1 |
Refseq |