Assay Details
| Gene Symbol: Seh1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039088.1 | NP_001034177.1 | 8 - 9 | 1204 | 112 | |
| GenBank mRNA | AK088835.1 | - | 8 - 9 | 1078 | 112 | |
| AK166573.1 | - | 8 - 9 | 1069 | 112 | ||
| AK169231.1 | - | 8 - 9 | 1085 | 112 | ||
| AK169388.1 | - | 8 - 9 | 1084 | 112 | ||
| AK170591.1 | - | 8 - 9 | 1157 | 112 | ||
| BC027244.1 | - | 8 - 9 | 1156 | 3584697 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Seh1l
Genomic Map