Assay Details
| Gene Symbol: Wnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271678.1 | NP_001258607.1 | 1 - 2 | 593 | 65 | |
| NM_001271679.1 | NP_001258608.1 | 1 - 2 | 593 | 65 | ||
| GenBank mRNA | JQ247189.1 | - | 1 - 2 | 593 | 65 | |
| JQ247190.1 | - | 1 - 2 | 593 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
positive regulation of sodium ion transport
positive regulation of peptidyl-threonine phosphorylation
phosphorylation
positive regulation of ion transmembrane transporter activity
negative regulation of apoptotic process
protein autophosphorylation
positive regulation of calcium ion transport
positive regulation of establishment of protein localization to plasma membrane
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
positive regulation of sodium ion transmembrane transporter activity
positive regulation of rubidium ion transport
positive regulation of rubidium ion transmembrane transporter activity
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
chloride channel inhibitor activity
protein phosphorylation
positive regulation of sodium ion transport
positive regulation of peptidyl-threonine phosphorylation
phosphorylation
positive regulation of ion transmembrane transporter activity
negative regulation of apoptotic process
protein autophosphorylation
positive regulation of calcium ion transport
positive regulation of establishment of protein localization to plasma membrane
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
positive regulation of sodium ion transmembrane transporter activity
positive regulation of rubidium ion transport
positive regulation of rubidium ion transmembrane transporter activity
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
chloride channel inhibitor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118392 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146403 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123864 | AK081752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123864 | NR_015600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123864 | BC043119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123864 | BC060731.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P810819 | AK034405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P194256 | BC043119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P194256 | BC060731.1 | mouse_transcriptome |
| Agilent Mouse cDNA Microarray (G4104A) | 804608 | AK002545.1 | genbank_mrna_mouse |
| Agilent Mouse cDNA Microarray (G4104A) | 804608 | AK167558.1 | genbank_mrna_mouse |
| Agilent Mouse cDNA Microarray (G4104A) | 804608 | AK171743.1 | genbank_mrna_mouse |
| Agilent Mouse cDNA Microarray (G4104A) | 804608 | BC145957.1 | genbank_mrna_mouse |
| Agilent Mouse cDNA Microarray (G4104A) | 804608 | NM_008326.1 | refseq_mrna_mouse_NM |
| Agilent Mouse cDNA Microarray (G4104A) | 804608 | U19119.1 | genbank_mrna_mouse |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15134906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15134909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14671810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14952378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15224245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14977086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15050767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10602452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10602448 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10602428 | - | - |
| Mouse Genome 430 2.0 Array | 1443924_at | - | - |
| Mouse Genome 430 2.0 Array | 1443924_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 279561_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 279561_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00009436.mm.1 | NM_001271678.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00009436.mm.1 | NM_001271679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00009436.mm.1 | TC0X00001514.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00009437.mm.1 | NM_001271678.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00009437.mm.1 | NM_001271679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00009437.mm.1 | TC0X00001514.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00009435.mm.1 | NM_001271678.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00009435.mm.1 | NM_001271679.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00009435.mm.1 | TC0X00001514.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443924_PM_at | - | - |
| Mouse Expression Set 430 | 1443924_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443924_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443924_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588691 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588691 | - | - |

Wnk3
Genomic Map