Assay Details
| Gene Symbol: Zyg11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167936.1 | NP_001161408.1 | 8 - 9 | 1539 | 77 | |
| GenBank mRNA | BC022150.1 | - | 6 - 7 | 1087 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zyg11a
Genomic Map