Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400015890.mm.1 |
NM_001085505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015890.mm.1 |
NM_001085505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015890.mm.1 |
TC0400001936.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015892.mm.1 |
NM_001085505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015892.mm.1 |
NM_001085505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015892.mm.1 |
TC0400001936.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015891.mm.1 |
NM_001085505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015891.mm.1 |
NM_001085505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015891.mm.1 |
TC0400001936.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015893.mm.1 |
NM_001085505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015893.mm.1 |
NM_001085505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015893.mm.1 |
TC0400001936.mm.1 |
MTA TC |