Assay Details
Gene Symbol: Gm572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085505.1 | NP_001078974.1 | 10 - 11 | 1119 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044182 | NM_001085505.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674962 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672771 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248399 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524417 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478206 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307334 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193566 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883418 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368542 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871045 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | AK156510.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | AK160326.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | BC021310.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | BC052905.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | NM_007651.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | X97227.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884267 | Z16071.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778458 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762502 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141840 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117111 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108152 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044182 | NM_001085505.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23395664 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03984358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395719 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03812702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03914222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04070956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03319390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03579200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395629 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03939534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395677 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03914227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03630296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03828587 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03666912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04008500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03778510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03984351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395690 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395823 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395834 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03240835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03984356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03432353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395685 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03432354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03939528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03780807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03630297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03837834 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395827 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04012651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04073362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395748 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03458575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395688 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03298833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03586465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03471512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395813 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03325250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03380745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395829 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03815058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395840 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03361866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03621986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23395657 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10510418 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510419 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510416 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510414 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510417 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510415 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10510413 | - | - |
Affymetrix Mouse Clariom S Array | PSR0400015894.mm.1 | NM_001085505.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400015894.mm.1 | NM_001085505.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400015894.mm.1 | TC0400001936.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400015896.mm.1 | NM_001085505.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400015896.mm.1 | NM_001085505.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400015896.mm.1 | TC0400001936.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400015895.mm.1 | NM_001085505.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400015895.mm.1 | NM_001085505.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400015895.mm.1 | TC0400001936.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 562853 | - | - |
