Assay Details
| Gene Symbol: Ldhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002257.1 | - | 3 - 4 | 316 | 68 | |
| AK019391.1 | - | 3 - 4 | 353 | 68 | ||
| AK131637.1 | - | 3 - 4 | 341 | 68 | ||
| BC046755.1 | - | 3 - 4 | 338 | 5693226 | 68 | |
| CX208147.1 | - | 3 - 4 | 327 | 68 | ||
| X51905.1 | - | 3 - 4 | 287 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ldhb
Genomic Map