Assay Details
| Gene Symbol: Ldhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002257.1 | - | 4 - 5 | 498 | 68 | |
| AK019391.1 | - | 4 - 5 | 535 | 68 | ||
| AK131637.1 | - | 4 - 5 | 523 | 68 | ||
| AK182230.1 | - | 1 - 2 | 77 | 68 | ||
| BC046755.1 | - | 4 - 5 | 520 | 5693226 | 68 | |
| CX208147.1 | - | 4 - 5 | 509 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ldhb
Genomic Map