Assay Details
| Gene Symbol: Tas1r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031872.2 | NP_114078.1 | 2 - 3 | 69 | ||
| GenBank mRNA | AB049994.1 | - | 2 - 3 | 69 | ||
| AF311386.1 | - | 2 - 3 | 69 | |||
| AF337039.1 | - | 2 - 3 | 69 | |||
| AY032621.1 | - | 2 - 3 | 492 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tas1r3
Genomic Map