Assay Details
| Gene Symbol: Hacd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025760.4 | NP_080036.1 | 4 - 5 | 434 | 112 | |
| GenBank mRNA | AK016968.1 | - | 4 - 5 | 418 | 112 | |
| AK017257.1 | - | 4 - 5 | 437 | 112 | ||
| AK039344.1 | - | 424 | 112 | |||
| AK039602.1 | - | 424 | 112 | |||
| AK048282.1 | - | 4 - 5 | 428 | 112 | ||
| AK050927.1 | - | 4 - 5 | 428 | 112 | ||
| BC020155.1 | - | 4 - 5 | 420 | 4016383 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hacd4
Genomic Map