Assay Details
Gene Symbol: BC049715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK044622.1 | - | 398 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK044622.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
