Assay Details
| Gene Symbol: Erp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026983.2 | NP_081259.1 | 5 - 6 | 634 | 55 | |
| GenBank mRNA | AK007684.1 | - | 5 - 6 | 635 | 55 | |
| AK131830.1 | - | 5 - 6 | 635 | 55 | ||
| BC037376.1 | - | 4 - 5 | 470 | 55 | ||
| BC104406.1 | - | 5 - 6 | 622 | 40050532 | 55 | |
| BC104407.2 | - | 5 - 6 | 622 | 40050533 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094292 | AK131830.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094292 | NM_026983.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094292 | AK007684.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682765 | AK032011.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78210 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179699 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172278 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094292 | AK131830.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094292 | NM_026983.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094292 | AK007684.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875234 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179699 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172278 | - | - |
| Whole Mouse Genome G4122A | A_52_P366696 | BC037376.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366696 | BC104406.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366696 | BC104407.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366696 | AK131830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366696 | NM_026983.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179699 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172278 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05776663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05800013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05523956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05776660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05342670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05024255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05180423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05006773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05006768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05006764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05435845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05024254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24875143 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548889 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452695_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430913_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548884 | - | - |
| Mouse Genome 430 2.0 Array | 1452695_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452695_at | - | - |
| Mouse Genome 430 2.0 Array | 1430913_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430913_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69187_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69187_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600026176.mm.1 | TC0600003336.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600026176.mm.1 | NM_026983.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600026176.mm.1 | NM_026983.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600026179.mm.1 | TC0600003336.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600026179.mm.1 | NM_026983.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600026179.mm.1 | NM_026983.2 | Refseq |
| Mouse Expression Set 430 | 1452695_PM_at | - | - |
| Mouse Expression Set 430 | 1452695_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430913_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452695_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430913_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452695_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 418903 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6380142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6380142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2658990 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2658990 | - | - |
| mouse-6 v1.1 (Illumina) | scl28273.7.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28273.7.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2888940 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2888940 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28273.7.1_5-S | - | - |

Erp27
Genomic Map