Assay Details
| Gene Symbol: Erp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC104406.1 | - | 6 - 7 | 823 | 40050532 | 68 |
| BC104407.2 | - | 6 - 7 | 823 | 40050533 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erp27
Genomic Map