Assay Details
| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029250.2 | NP_083526.2 | 3 - 4 | 811 | 80 | |
| GenBank mRNA | AK016135.1 | - | 3 - 4 | 793 | 80 | |
| AK031560.1 | - | 788 | 80 | |||
| AK033268.1 | - | 3 - 4 | 788 | 80 | ||
| AK044502.1 | - | 3 - 4 | 812 | 80 | ||
| AK049827.1 | - | 3 - 4 | 525 | 80 | ||
| AK051039.1 | - | 821 | 80 | |||
| AK164928.1 | - | 3 - 4 | 788 | 80 | ||
More Information
Set Membership: |
|

Etnk1
Genomic Map