Assay Details
| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029250.2 | NP_083526.2 | 7 - 8 | 1268 | 93 | |
| GenBank mRNA | AK016135.1 | - | 7 - 8 | 1250 | 93 | |
| AK033268.1 | - | 7 - 8 | 1245 | 93 | ||
| AK044502.1 | - | 7 - 8 | 1269 | 93 | ||
| AK049827.1 | - | 7 - 8 | 982 | 93 | ||
More Information
Set Membership: |
|

Etnk1
Genomic Map