Assay Details
| Gene Symbol: Slc24a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110240.1 | NP_001103710.1 | 1 - 2 | 1080 | 59 | |
| GenBank mRNA | AK034424.1 | - | 2 - 3 | 1189 | 59 | |
| AK038800.1 | - | 2 - 3 | 1233 | 59 | ||
| BC115867.1 | - | 1 - 2 | 1082 | 40081382 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc24a2
Genomic Map