Assay Details
| Gene Symbol: Slc24a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172426.2 | NP_766014.1 | 5 - 6 | 1504 | 98 | |
| GenBank mRNA | AK012771.1 | - | 310 | 98 | ||
| AK083127.1 | - | 5 - 6 | 1388 | 98 | ||
More Information
Set Membership: |
|

Slc24a2
Genomic Map