Assay Details
| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177772.1 | NP_001171243.1 | 9 - 10 | 1579 | 153 | |
| NM_021471.2 | NP_067446.1 | 9 - 10 | 1579 | 153 | ||
| GenBank mRNA | AK048926.1 | - | 9 - 10 | 1549 | 153 | |
| AY007379.1 | - | 9 - 10 | 1415 | 153 | ||
| BC078456.1 | - | 9 - 10 | 1529 | 30360663 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slco1c1
Genomic Map