Assay Details
| Gene Symbol: 1700060C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045732.1 | 2 - 3 | 164 | 70 | ||
| GenBank mRNA | AK006838.1 | - | 2 - 3 | 164 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930579D09 gene | 4930579D09Rik | -79.9kb | Sense | Chr. 6 - 143533943 - 143535533 on NCBI Build GRCm38 | |
| Downstream | SRY (sex determining region Y)-box 5 | Sox5 |
A730017D01Rik, AI528773 |
193.5kb | Sense | Chr. 6 - 143828425 - 144781989 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700060C16Rik
Genomic Map