Assay Details
| Gene Symbol: Megf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172694.2 | NP_766282.1 | 2 - 3 | 910 | 60 | |
| GenBank mRNA | AK035987.1 | - | 2 - 3 | 911 | 60 | |
| AK039319.1 | - | 2 - 3 | 911 | 60 | ||
| AK122378.1 | - | 901 | 60 | |||
| BC088994.1 | - | 2 - 3 | 819 | 6734479 | 60 | |
| BC138128.1 | - | 2 - 3 | 849 | 8861147 | 60 | |
| BC138129.1 | - | 2 - 3 | 849 | 8861148 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Megf9
Genomic Map