Assay Details
| Gene Symbol: Pole2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011133.2 | NP_035263.1 | 5 - 6 | 445 | 97 | |
| GenBank mRNA | AF036898.1 | - | 5 - 6 | 418 | 97 | |
| AK077592.1 | - | 5 - 6 | 446 | 97 | ||
| AK172024.1 | - | 5 - 6 | 440 | 97 | ||
| BC052170.1 | - | 5 - 6 | 434 | 97 | ||
| BC063772.1 | - | 5 - 6 | 434 | 30147626 | 97 | |
More Information
Set Membership: |
|

Pole2
Genomic Map