Assay Details
| Gene Symbol: Sh3yl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013709.5 | NP_038737.1 | 3 - 4 | 785 | 58 | |
| NR_110343.1 | 4 - 5 | 930 | 58 | |||
| GenBank mRNA | AK141199.1 | - | 3 - 4 | 786 | 58 | |
| AK168483.1 | - | 4 - 5 | 413 | 58 | ||
| BC010296.1 | - | 2 - 3 | 233 | 3154966 | 58 | |
| BC061488.1 | - | 3 - 4 | 725 | 4483634 | 58 | |
| BC110043.1 | - | 3 - 4 | 763 | 5149633 | 58 | |
| D85926.1 | - | 3 - 4 | 268 | 58 | ||
More Information
Set Membership: |
|

Sh3yl1
Genomic Map