Assay Details
| Gene Symbol: Sh3yl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289480.1 | NP_001276409.1 | 4 - 5 | 845 | 78 | |
| NM_013709.5 | NP_038737.1 | 5 - 6 | 959 | 78 | ||
| NR_110343.1 | 6 - 7 | 1104 | 78 | |||
| GenBank mRNA | AK141199.1 | - | 5 - 6 | 960 | 78 | |
| BC010296.1 | - | 4 - 5 | 407 | 3154966 | 78 | |
| BC058557.1 | - | 4 - 5 | 822 | 6313231 | 78 | |
| BC061488.1 | - | 5 - 6 | 899 | 4483634 | 78 | |
| BC110043.1 | - | 5 - 6 | 937 | 5149633 | 78 | |
| D85926.1 | - | 5 - 6 | 442 | 78 | ||
More Information
Set Membership: |
|

Sh3yl1
Genomic Map