Assay Details
| Gene Symbol: Sh3yl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289480.1 | NP_001276409.1 | 6 - 7 | 1133 | 79 | |
| NM_013709.5 | NP_038737.1 | 7 - 8 | 1247 | 79 | ||
| NR_110343.1 | 8 - 9 | 1392 | 79 | |||
| GenBank mRNA | AK141199.1 | - | 7 - 8 | 1248 | 79 | |
| AK168483.1 | - | 8 - 9 | 873 | 79 | ||
| BC010296.1 | - | 6 - 7 | 695 | 3154966 | 79 | |
| BC058557.1 | - | 6 - 7 | 1110 | 6313231 | 79 | |
| BC110043.1 | - | 7 - 8 | 1225 | 5149633 | 79 | |
| D85926.1 | - | 7 - 8 | 730 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3yl1
Genomic Map