Assay Details
| Gene Symbol: Sh3yl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289480.1 | NP_001276409.1 | 7 - 8 | 1220 | 146 | |
| NM_013709.5 | NP_038737.1 | 8 - 9 | 1334 | 146 | ||
| NR_110343.1 | 9 - 10 | 1479 | 146 | |||
| GenBank mRNA | AK141199.1 | - | 8 - 9 | 1335 | 146 | |
| AK168483.1 | - | 9 - 10 | 960 | 146 | ||
| BC010296.1 | - | 7 - 8 | 782 | 3154966 | 146 | |
| BC058557.1 | - | 7 - 8 | 1197 | 6313231 | 146 | |
| BC061488.1 | - | 7 - 8 | 1934 | 4483634 | 146 | |
| BC110043.1 | - | 8 - 9 | 1312 | 5149633 | 146 | |
| D85926.1 | - | 8 - 9 | 817 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3yl1
Genomic Map