Assay Details
| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093775.1 | NP_001087244.1 | 6 - 7 | 961 | 75 | |
| NM_001093776.1 | NP_001087245.1 | 6 - 7 | 873 | 75 | ||
| NM_001093778.1 | NP_001087247.1 | 6 - 7 | 961 | 75 | ||
| NM_008666.3 | NP_032692.2 | 6 - 7 | 961 | 75 | ||
| GenBank mRNA | AB093283.1 | - | 3 - 4 | 320 | 75 | |
| AB212898.1 | - | 5 - 6 | 559 | 75 | ||
| AF004295.1 | - | 6 - 7 | 786 | 72 | ||
| AK049624.1 | - | 6 - 7 | 661 | 75 | ||
| BC079651.1 | - | 6 - 7 | 842 | 75 | ||
| BC094438.1 | - | 2 - 3 | 198 | 30692780 | 75 | |
| BC131677.1 | - | 5 - 6 | 562 | 40111036 | 75 | |
| U86338.1 | - | 6 - 7 | 680 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myt1l
Genomic Map