Assay Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015099.2 | NP_001015099.2 | 2 - 3 | 256 | 97 | |
| NM_001167963.1 | NP_001161435.1 | 2 - 3 | 256 | 97 | ||
| NM_001167964.1 | NP_001161436.1 | 2 - 3 | 256 | 97 | ||
| GenBank mRNA | AK031333.1 | - | 2 - 3 | 116 | 97 | |
| AK033513.1 | - | 2 - 3 | 198 | 97 | ||
| AK084256.1 | - | 2 - 3 | 127 | 97 | ||
| AK129332.1 | - | 2 - 3 | 159 | 97 | ||
| BC086455.1 | - | 2 - 3 | 129 | 30458160 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084256.1, BC086455.1, NM_001015099.2, NM_001167963.1, NM_001167964.1 transcripts.
Set Membership: |
|

G2e3
Genomic Map