Assay Details
| Gene Symbol: Dhx34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285931.1 | NP_001272860.1 | 10 - 11 | 3106 | 54 | |
| NM_001285932.1 | NP_001272861.1 | 10 - 11 | 2872 | 54 | ||
| NM_027883.3 | NP_082159.3 | 10 - 11 | 2597 | 54 | ||
| GenBank mRNA | AK004733.1 | - | 10 - 11 | 2598 | 54 | |
| AK007461.1 | - | 1 - 2 | 339 | 54 | ||
| AK036763.1 | - | 6 - 7 | 998 | 54 | ||
| AK076217.1 | - | 9 - 10 | 2573 | 54 | ||
| AK129062.1 | - | 10 - 11 | 2580 | 54 | ||
| BC058370.1 | - | 10 - 11 | 2934 | 54 | ||
| BC080856.1 | - | 10 - 11 | 2872 | 6854931 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007461.1 transcripts.
Set Membership: |
|

Dhx34
Genomic Map