Assay Details
| Gene Symbol: Dhx34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285931.1 | NP_001272860.1 | 7 - 8 | 2571 | 58 | |
| NM_001285932.1 | NP_001272861.1 | 7 - 8 | 2337 | 58 | ||
| NM_027883.3 | NP_082159.3 | 7 - 8 | 2062 | 58 | ||
| GenBank mRNA | AK004733.1 | - | 7 - 8 | 2063 | 58 | |
| AK036763.1 | - | 3 - 4 | 463 | 58 | ||
| AK076217.1 | - | 6 - 7 | 2038 | 58 | ||
| AK129062.1 | - | 7 - 8 | 2045 | 58 | ||
| AK177329.1 | - | 2 - 3 | 209 | 58 | ||
| BC058370.1 | - | 7 - 8 | 2399 | 58 | ||
| BC080856.1 | - | 7 - 8 | 2337 | 6854931 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx34
Genomic Map