Assay Details
| Gene Symbol: Dhx34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285931.1 | NP_001272860.1 | 9 - 10 | 2868 | 69 | |
| NM_001285932.1 | NP_001272861.1 | 9 - 10 | 2634 | 69 | ||
| NM_027883.3 | NP_082159.3 | 9 - 10 | 2359 | 69 | ||
| GenBank mRNA | AK004733.1 | - | 9 - 10 | 2360 | 69 | |
| AK036763.1 | - | 5 - 6 | 760 | 69 | ||
| AK076217.1 | - | 8 - 9 | 2335 | 69 | ||
| AK129062.1 | - | 9 - 10 | 2342 | 69 | ||
| BC058370.1 | - | 9 - 10 | 2696 | 69 | ||
| BC080856.1 | - | 9 - 10 | 2634 | 6854931 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx34
Genomic Map