Assay Details
| Gene Symbol: Strn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039878.2 | NP_001034967.1 | 3 - 4 | 929 | 116 | |
| NM_133789.3 | NP_598550.2 | 3 - 4 | 929 | 116 | ||
| GenBank mRNA | AF414080.1 | - | 3 - 4 | 929 | 116 | |
| AK136155.1 | - | 3 - 4 | 245 | 116 | ||
| AK154615.1 | - | 3 - 4 | 479 | 116 | ||
| AK161673.1 | - | 3 - 4 | 501 | 116 | ||
| BC080283.1 | - | 3 - 4 | 495 | 6401782 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strn4
Genomic Map