Assay Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019426.2 | NP_062299.2 | 11 - 12 | 3171 | 66 | |
| GenBank mRNA | AK132359.1 | - | 10 - 11 | 1791 | 66 | |
| AK137199.1 | - | 3 - 4 | 330 | 66 | ||
| AK167542.1 | - | 12 - 13 | 3217 | 66 | ||
| BC052460.1 | - | 11 - 12 | 3171 | 6834550 | 66 | |
| BC060246.1 | - | 11 - 12 | 3154 | 5705945 | 66 | |
More Information
Set Membership: |
|

Atf7ip
Genomic Map