Assay Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019426.2 | NP_062299.2 | 13 - 14 | 3508 | 88 | |
| GenBank mRNA | AJ132702.1 | - | 13 - 14 | 3501 | 88 | |
| AK132359.1 | - | 12 - 13 | 2128 | 88 | ||
| AK137199.1 | - | 5 - 6 | 667 | 88 | ||
| AK157479.1 | - | 3 - 4 | 1267 | 88 | ||
| AK167542.1 | - | 14 - 15 | 3554 | 88 | ||
| BC052460.1 | - | 13 - 14 | 3508 | 6834550 | 88 | |
| BC060246.1 | - | 13 - 14 | 3491 | 5705945 | 88 | |
More Information
Set Membership: |
|

Atf7ip
Genomic Map