Assay Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019426.2 | NP_062299.2 | 6 - 7 | 2217 | 109 | |
| GenBank mRNA | AJ132702.1 | - | 6 - 7 | 2210 | 109 | |
| AK132359.1 | - | 5 - 6 | 837 | 109 | ||
| AK167542.1 | - | 7 - 8 | 2263 | 109 | ||
| BC037617.1 | - | 2219 | 109 | |||
| BC052460.1 | - | 6 - 7 | 2217 | 6834550 | 109 | |
| BC060246.1 | - | 6 - 7 | 2200 | 5705945 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atf7ip
Genomic Map