Assay Details
| Gene Symbol: Csnk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027874.2 | NP_082150.1 | 9 - 10 | 1583 | 76 | |
| GenBank mRNA | AK004606.1 | - | 9 - 10 | 1343 | 76 | |
| AK146600.1 | - | 9 - 10 | 1573 | 76 | ||
| AK150387.1 | - | 9 - 10 | 1312 | 76 | ||
| AK150815.1 | - | 9 - 10 | 1310 | 76 | ||
More Information
Set Membership: |
|

Csnk1d
Genomic Map