Assay Details
| Gene Symbol: Aspscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026877.2 | NP_081153.1 | 11 - 12 | 1389 | 69 | |
| NM_198223.2 | NP_937866.1 | 10 - 11 | 1168 | 69 | ||
| GenBank mRNA | AK004509.1 | - | 5 - 6 | 401 | 69 | |
| AK088197.1 | - | 11 - 12 | 1227 | 69 | ||
| AY349132.1 | - | 9 - 10 | 1158 | 69 | ||
| AY349133.1 | - | 11 - 12 | 1306 | 69 | ||
| BC019177.1 | - | 11 - 12 | 1332 | 5008127 | 69 | |
| BC022115.1 | - | 11 - 12 | 1336 | 5101238 | 69 | |
| BC031153.1 | - | 11 - 12 | 1321 | 4976569 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aspscr1
Genomic Map