Assay Details
| Gene Symbol: Errfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133753.1 | NP_598514.1 | 2 - 3 | 368 | 144 | |
| GenBank mRNA | AK004851.1 | - | 2 - 3 | 387 | 144 | |
| AK139780.1 | - | 2 - 3 | 384 | 144 | ||
| AK166791.1 | - | 2 - 3 | 387 | 144 | ||
| AK167156.1 | - | 2 - 3 | 327 | 144 | ||
| AK167359.1 | - | 2 - 3 | 387 | 144 | ||
| AK205245.1 | - | 1 - 2 | 199 | 144 | ||
| AK212670.1 | - | 1 - 2 | 219 | 144 | ||
| BC005546.1 | - | 2 - 3 | 368 | 3585640 | 144 | |
| BC057646.1 | - | 2 - 3 | 386 | 4194124 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Errfi1
Genomic Map