Assay Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027238.2 | NP_081514.1 | 12 - 13 | 1055 | 66 | |
| GenBank mRNA | AK037208.1 | - | 12 - 13 | 1056 | 66 | |
| AK049108.1 | - | 12 - 13 | 1106 | 66 | ||
| AK132655.1 | - | 7 - 8 | 506 | 66 | ||
| BC151021.1 | - | 12 - 13 | 1017 | 9087934 | 66 | |
More Information
Set Membership: |
|

Ttc39b
Genomic Map