Assay Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027238.2 | NP_081514.1 | 2 - 3 | 152 | 71 | |
| GenBank mRNA | AK007856.1 | - | 2 - 3 | 153 | 71 | |
| AK037208.1 | - | 2 - 3 | 153 | 71 | ||
| AK049108.1 | - | 2 - 3 | 203 | 71 | ||
| BC151021.1 | - | 2 - 3 | 114 | 9087934 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39b
Genomic Map