Assay Details
| Gene Symbol: Mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010773.2 | NP_034903.2 | 2 - 3 | 838 | 102 | |
| GenBank mRNA | AF072243.1 | - | 2 - 3 | 796 | 102 | |
| AK147008.1 | - | 2 - 3 | 723 | 102 | ||
| BC026698.1 | - | 2 - 3 | 798 | 102 | ||
| BC046607.1 | - | 2 - 3 | 838 | 6399206 | 102 | |
| BC057289.1 | - | 2 - 3 | 670 | 102 | ||
| BC058864.1 | - | 2 - 3 | 658 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbd2
Genomic Map