Assay Details
| Gene Symbol: Lipg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010720.3 | NP_034850.3 | 4 - 5 | 857 | 91 | |
| GenBank mRNA | AF118768.1 | - | 4 - 5 | 842 | 91 | |
| AK090224.1 | - | 4 - 5 | 856 | 91 | ||
| AK152161.1 | - | 4 - 5 | 859 | 91 | ||
| AK152430.1 | - | 4 - 5 | 855 | 91 | ||
| AK153541.1 | - | 4 - 5 | 859 | 91 | ||
| AK172101.1 | - | 4 - 5 | 857 | 91 | ||
| BC020991.1 | - | 4 - 5 | 711 | 3981856 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lipg
Genomic Map