Assay Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291019.1 | NP_001277948.1 | 2 - 3 | 884 | 115 | |
| NM_001291020.1 | NP_001277949.1 | 3 - 4 | 686 | 115 | ||
| NM_007547.4 | NP_031573.2 | 2 - 3 | 884 | 115 | ||
| GenBank mRNA | AB018194.1 | - | 2 - 3 | 450 | 115 | |
| AF072543.1 | - | 2 - 3 | 440 | 115 | ||
| AF332080.1 | - | 2 - 3 | 450 | 115 | ||
| AK159617.1 | - | 3 - 4 | 689 | 115 | ||
| BC062197.1 | - | 2 - 3 | 475 | 5368250 | 115 | |
More Information
Set Membership: |
|

Sirpa
Genomic Map