Assay Details
| Gene Symbol: Tgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009374.3 | NP_033400.2 | 7 - 8 | 1048 | 61 | |
| GenBank mRNA | BC129890.1 | - | 7 - 8 | 1014 | 40111487 | 61 |
| BC129891.1 | - | 7 - 8 | 1014 | 40111493 | 61 | |
| L10385.1 | - | 7 - 8 | 1039 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgm3
Genomic Map