Assay Details
| Gene Symbol: Mmp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152944.1 | NP_694423.1 | 2 - 3 | 708 | 79 | |
| GenBank mRNA | AF507967.1 | - | 2 - 3 | 693 | 79 | |
| AY124569.1 | - | 2 - 3 | 708 | 79 | ||
| BC106860.2 | - | 2 - 3 | 708 | 40047272 | 79 | |
| BC106861.2 | - | 2 - 3 | 708 | 40047279 | 79 | |
| BC111522.1 | - | 1 - 2 | 1515 | 40047286 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp21
Genomic Map