Assay Details
| Gene Symbol: Mmp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152944.1 | NP_694423.1 | 6 - 7 | 1417 | 75 | |
| GenBank mRNA | AF507967.1 | - | 6 - 7 | 1402 | 75 | |
| AY124569.1 | - | 6 - 7 | 1417 | 75 | ||
| BC106861.2 | - | 6 - 7 | 1417 | 40047279 | 75 | |
| BC111522.1 | - | 5 - 6 | 2224 | 40047286 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170841 | BC106860.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170841 | BC106861.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170841 | BC111522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170841 | NM_152944.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170841 | AY124569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046985 | NM_152944.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046985 | AY124569.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285641 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170841 | BC106860.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170841 | BC106861.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170841 | BC111522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170841 | NM_152944.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170841 | AY124569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046985 | NM_152944.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046985 | AY124569.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272866 | BC106860.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272866 | BC106861.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272866 | BC111522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272866 | NM_152944.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272866 | AF507967.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25541997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25542011 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01642026 | - | - |
| Whole Mouse Genome G4122A | A_51_P272866 | BC106860.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272866 | BC106861.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272866 | BC111522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272866 | NM_152944.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272866 | AF507967.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115739 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139345 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06529140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05930475 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05930478 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05930474 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05930481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06529137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25542005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25541960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06647891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06647882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05930481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06410717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06279538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06029565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06410711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06112275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06029552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25542028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06616126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25541982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568631 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568629 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700037182.mm.1 | NM_152944.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037182.mm.1 | NM_152944.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037182.mm.1 | TC0700004427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700037181.mm.1 | NM_152944.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037181.mm.1 | NM_152944.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037181.mm.1 | TC0700004427.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 802355 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4480673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223199 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223199 | - | - |
| mouse-6 v1.1 (Illumina) | scl30563.7.1_113-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30563.7.1_113-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223199 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223199 | - | - |

Mmp21
Genomic Map